Sodium formate ≥99%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:141-53-7
:
Formula:HCO₂Na
MDL Number:MFCD00013101
:68.01 g/mol
Merck Index:13,08694
Melting Pt:257.31 °C
Density:1.919 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-488-0
REACH:01-2119486468-21
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 10 ppm
- Ca (Calcium):Max. 50 ppm
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84850.260 spec_VWRC84850.260 spec_VWRC84850.460