D-(+)-Xylose
Catalog # ENAMEN300107152.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.13 g/mol
Merck Index:13,10142
Boiling Pt:331 °C (1013 hPa)
Melting Pt:154 °C
Density:1.525 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-400-7
CAS Number:58-86-6
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475