Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenol liquified
Phenol liquified
Phenol liquified
Phenol liquified
Supplier:
Uptima
Certificates
Expand image
Phenol liquified
Supplier:
Uptima
About this item
CAS Number:
108-95-2
:
Formula:
C₆H₅OH
MDL Number:
MFCD00002143
:
94.11 g/mol
Storage Temperature:
Ambient
EINECS:
203-632-7
UN:
2821
ADR:
6.1,II
:
GHS06
:
GHS05
:
GHS08