Bisacrylamide
ENAMEN300104853.1G
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
:
:154.17 g/mol
:125 °C (3.33 hPa)
:185 °C
:1.235 g/cm³ (20 °C)
:138 °C
:Refrigerator
:203-750-9
:GHS07
:110-26-9
:(H₂C=CHCONH)₂CH₂
:MFCD00008625