1-(3-Aminopropyl)piperidine
Catalog # ABCRMK362.0010
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:142.2419 g/mol
EINECS:222-557-0
CAS Number:3529-08-6
Formula:C₈H₁₈N₂
MDL Number:MFCD00023784