Order Entry
Germany
ContactUsLinkComponent
2,3-Dichloro-5-(trichloromethyl)pyridine
2,3-Dichloro-5-(trichloromethyl)pyridine
Catalog # MOLEM45141459-5G
Supplier:  Molekula
CAS Number:  
2,3-Dichloro-5-(trichloromethyl)pyridine
Catalog # MOLEM45141459-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :265.3518 g/mol
CAS Number:69045-83-6
Formula:C₆H₂Cl₅N
MDL Number:MFCD03701543