0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Thymolphthaleine
Thymolphthalein 1 g/l in denatured ethanol
Thymolphthalein 1 g/l in denatured ethanol
Thymolphthalein 1 g/l in denatured ethanol
Supplier:
Bernd Kraft
Certificates
Expand image
Thymolphthalein 1 g/l in denatured ethanol
Supplier:
Bernd Kraft
Certificates
About this item
CAS Number:
125-20-2
:
Formula:
C₂₈H₃₀O₄
MDL Number:
MFCD00005909
:
430.54 g/mol
Storage Temperature:
Ambient
EINECS:
204-729-7
UN:
1170
ADR:
3,II
:
GHS02
Related Information
Documentation
Certificates
Search by Lot Number
Documentation