Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
Supplier:
COMBI-BLOCKS
Certificates
Expand image
4-(2,2,2-Trifluoroethoxy)phenylboronic acid
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
886536-37-4
:
Formula:
C₈H₈BF₃O₃
MDL Number:
MFCD09027239
:
219.95 g/mol