
2-Formyl-6-picoline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1122-72-1
:
Formula:C₇H₇NO
MDL Number:MFCD00006291
:121.14 g/mol
Melting Pt:31…33 °C
Flash Pt:68 °C
EINECS:214-359-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- HPLC:≥96.0 %