Ethyl 4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylate
Catalog # COBBHC-6334-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.22 g/mol
CAS Number:71083-04-0
Formula:C₁₃H₁₀F₃NO₃
MDL Number:MFCD01080777