2,3,5-Triphenyltetrazolium chloride (TTC) 1% in aqueous solution
Catalog # HACH2406042
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:334.81 g/mol
EINECS:206-071-6
CAS Number:298-96-4
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963