Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-33-7
:
Formula:W
MDL Number:MFCD00011461
Storage Temperature:Ambient
EINECS:231-143-9
Frequently Bought Together