Buffers, carbonate, 0,5 M solutions
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 497-19-8
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00003494
8 Options Available Below
Product Details & Documents
CAS No.: 497-19-8
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00003494
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J63899.AK
- J63899.AP
- J63658.AK
- J63658.AP
- J62610.AK
- J62610.AP
- J61229.AK
- J61229.AP
- Odour/scent:
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Physical form:
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- CAS No.:
- 497-19-8
- 497-19-8
- 497-19-8
- 497-19-8
- 497-19-8
- 497-19-8
- 497-19-8
- 497-19-8
- MDL No.:
- MFCD00003494
- MFCD00003494
- MFCD00003494
- MFCD00003494
- MFCD00003494
- MFCD00003494
- MFCD00003494
- MFCD00003494
Specifications
Product Family Options
Product Information
- ConcentrationpH valuePack typePkAvailabilityPrice
- J63899.AKCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J63899.AKOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J63899.APCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J63899.APOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J63658.AKCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J63658.AKOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J63658.APCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J63658.APOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J62610.AKCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J62610.AKOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J62610.APCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J62610.APOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J61229.AKCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J61229.AKOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494 - J61229.APCarbonate buffer solutionSpecifications:
More
Specifications
Cat. No.J61229.APOdour/scentOdourlessPhysical formLiquidCAS No.497-19-8MDL No.MFCD00003494
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



