Isoquinoline-4-carbaldehyde
Catalog # ENAMEN300100667.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.17 g/mol
CAS Number:22960-16-3
Formula:C₁₀H₇NO
MDL Number:MFCD00829440