4-Cyano-2-fluorobenzaldehyde
Catalog # ENAMEN300107978.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:149.1200 g/mol
CAS Number:105942-10-7
Formula:C₈H₄FNO
MDL Number:MFCD07368786