Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Chloro-2'-hydroxypropiophenone

5'-Chloro-2'-hydroxypropiophenone

1 g
Catalog # ENAMEN30010779.1G
Marketsource
Supplier:   SIA ENAMINE
5'-Chloro-2'-hydroxypropiophenone
Catalog # ENAMEN30010779.1G
Supplier:  SIA ENAMINE
Marketsource
CAS Number:  2892-16-2
5'-Chloro-2'-hydroxypropiophenone

1 g

CAS Number:  2892-16-2
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:2892-16-2
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00218579
Specifications

Specifications

  • Pk:
    1 g
  • Cat. No.:
    ENAMEN30010779.1G

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.