Benzo[b]thiophene-2-carboxaldehyde
Catalog # ENAMEN300105457.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.21 g/mol
CAS Number:3541-37-5
Formula:C₉H₆OS
MDL Number:MFCD01075041