Tubing sleeves, PEEK, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
PEEK, natural or coloured.
The PEEK tubing sleeve facilitates easy connection of capillary tubing to standard, coned ports. Manufactured from biocompatible PEEK it offers optimal chemical resistance. By using a sleeve instead of a modified ferrule, dead volume in the receiving port is minimized, ensuring better results.