Occupational shoes, clogs, Chiro Clogs Classic
Supplier: SCHURR SCHUHVERTRIEB
Certificates
About this item
The anti-static Chiro Clogs are made from PU.
- Wash- and disinfectable
- PU fleece lining
- Excellent shock absorption in heel area
- Ergonomic heel hight
- Anatomically shaped integrated insole
- Without heel strap
For use in high-sensitive areas like hospitals, hygiene areas, pharmaceutical industries and research laboratories.
Colours: White, green, blue, red, yellow
Certifications: EN ISO 20347:2004