Orders LinkContactUsLinkComponent
Safety helmets, Reflex
Safety helmets, Reflex
Certificates
Safety helmets, Reflex
Safety helmets, Reflex
56 Options Available Below
Product Details & Documents
These vented safety helmets are made of HDPE. They are available with or without wheel ratchet.
  • 6-point nylon textile cradle, LDPE headband
  • Sweatband made of brushed nylon with 2 mm foam
  • High visibility options for improved visibility during poor light conditions
  • Compatible with the CONNECT and EURO face and hearing accessories
Size: 51 to 63 cm (standard); 53 to 64 cm (with ratchet)
Weight: 350 g (standard); 370 g (with ratchet)
 : EN 397:1995+A1 / EN 397:2012+A1:2012; EN 50365:2002
Product Family Options
Vented, with silver flash
Product Information
  • Colour
    Availability
    Price
  • CENUS12KSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12BSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12GSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12ESA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12OSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12RSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12WSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12YSA
    Safety helmet, vented, with silver flash, without ratchet
    Safety helmet, vented, with silver flash, without ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
Vented, with silver flash and ratchet
Product Information
  • Colour
    Availability
    Price
  • CENUS12KSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12BSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12GSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12ESR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12OSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12RSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12WSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12YSR
    Safety helmet, vented, with silver flash and ratchet
    Safety helmet, vented, with silver flash and ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
Vented, with high visibility flash
Product Information
  • Colour
    Availability
    Price
  • CENUS12KHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12KHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12BHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12BHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12GHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12GHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12EHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12EHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12OHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12OHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12RHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12RHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12WHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12WHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12YHVOA
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Safety helmet, vented, with high visibility orange flash, without ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • CENUS12YHVYA
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Safety helmet, vented, with high visibility yellow flash, without ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
Vented, with high visibility flash and ratchet
Product Information
  • Colour
    Availability
    Price
  • CENUS12KHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12KHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12BHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12BHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12GHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12GHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12EHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12EHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12OHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12OHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12RHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12RHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12WHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12WHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12YHVOR
    Safety helmet, vented, with high visibility orange flash and ratchet
    Safety helmet, vented, with high visibility orange flash and ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
  • CENUS12YHVYR
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Safety helmet, vented, with high visibility yellow flash and ratchet
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,70 €
    23,70 €
    1 * 1 ST
       23,70 €
Complete high visibility helmets, vented, without ratchet
Product Information
  • Colour
    Availability
    Price
  • CENUS12HVOHVOA
    Safety high visibility helmet, complete, vented, with high visibility orange flash, without ratchet
    Safety high visibility helmet, complete, vented, with high visibility orange flash, without ratchet
    Hi-vis orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • CENUS12HVOHVYA
    Safety high visibility helmet, complete, vented, with high visibility yellow flash, without ratchet
    Safety high visibility helmet, complete, vented, with high visibility yellow flash, without ratchet
    Hi-vis orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • CENUS12HVYHVOA
    Safety high visibility helmet, complete, vented, with high visibility orange flash, without ratchet
    Safety high visibility helmet, complete, vented, with high visibility orange flash, without ratchet
    Hi-vis yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • CENUS12HVYHVYA
    Safety high visibility helmet, complete, vented, with high visibility yellow flash, without ratchet
    Safety high visibility helmet, complete, vented, with high visibility yellow flash, without ratchet
    Hi-vis yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
Complete high visibility helmets, vented, with ratchet
Product Information
  • Colour
    Availability
    Price
  • CENUS12HVOHVOR
    Safety high visibility helmet, complete, vented, with high visibility orange flash and ratchet
    Safety high visibility helmet, complete, vented, with high visibility orange flash and ratchet
    Hi-vis orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €
  • CENUS12HVOHVYR
    Safety high visibility helmet, complete, vented, with high visibility yellow flash and ratchet
    Safety high visibility helmet, complete, vented, with high visibility yellow flash and ratchet
    Hi-vis orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €
  • CENUS12HVYHVOR
    Safety high visibility helmet, complete, vented, with high visibility orange flash and ratchet
    Safety high visibility helmet, complete, vented, with high visibility orange flash and ratchet
    Hi-vis yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €
  • CENUS12HVYHVYR
    Safety high visibility helmet, complete, vented, with high visibility yellow flash and ratchet
    Safety high visibility helmet, complete, vented, with high visibility yellow flash and ratchet
    Hi-vis yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €