VWR® PCR DNA Marker
E854-100RXN
:
- Pk:1 KIT
- Description:PCR DNA Marker™
PCR DNA Marker™ offers 8 fragments ranging from 50 to 2000 bp. The marker contains high intensity reference bands that are ideal for quick sizing of PCR products and restriction digests. Simply mix 10 μl of PCR DNA Marker™ with required quantity of loading buffer and load into well. There is sufficient material for 100 assays.
- High resolution marker optimal for size estimation of PCR fragments