VWR® DNA Molecular Weight Marker, High-Range
Catalog # E255-150UG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:150 µG
- Description:DNA MW marker, High-Range
Specifications
About this item
High-Range DNA MW (molecular weight) marker offers 10 fragments ranging from 1503 to 48 502 bp. There is sufficient material for 75 to 150 assays.
- Optimal for evaluation fragments >10 kb
- Easy band identification