Hydrogen peroxide 5%, Technical Grade for desinfection of tube systems
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Density:1.022 g/cm³ (20 °C)
:34.01 g/mol
EINECS:231-765-0
:GHS05
Storage Temperature:Ambient