Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrogen peroxide
Hydrogen peroxide 12%, extra pure
Hydrogen peroxide 12%, extra pure
Hydrogen peroxide 12%, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Hydrogen peroxide 12%, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
7722-84-1
:
Formula:
H₂O₂
MDL Number:
MFCD00011333
ADR:
5.1,III
:
34.01 g/mol
EINECS:
231-765-0
UN:
2984
:
GHS05
Storage Temperature:
Ambient