Acetic acid ≥98%, tested according to Ph. Eur., Fluka™
Catalog # 45741-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Explosion Limit:16 %, 92 °F
- Identity A:complying
- Identity B:complying
- Appearance of the substance:complying
- Assay:99.0 - 100.5 %
- Non-volatile matter:Max. 0.01 %
- Congealing - Freezing Point:Min. 14.8 °C
- Iron (Fe):Max. 5 ppm
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 23 ppm
- Sulfate (SO₄):Max. 47 ppm
- Reducing impurities:complying
- Residual solvents:complying
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152