Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium thiosulphate
Sodium thiosulphate 0.002 mol/l (0.002 N) in aqueous solution
Sodium thiosulphate 0.002 mol/l (0.002 N) in aqueous solution
Sodium thiosulphate 0.002 mol/l (0.002 N) in aqueous solution
KRAF13935.3000
:
Bernd Kraft
:
Sodium thiosulphate 0.002 mol/l (0.002 N) in aqueous solution
KRAF13935.3000
:
Bernd Kraft
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499