0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Hydrogen peroxide
Hydrogen peroxide 30% analytical reagent
Hydrogen peroxide 30% analytical reagent
:
Bernd Kraft
Hydrogen peroxide 30% analytical reagent
:
Bernd Kraft
:
7722-84-1
:
:
H₂O₂
:
MFCD00011333
:
∼107 °C (1013 hPa)
:
< 0 °C
:
1.11 g/cm³ (20 °C)
:
231-765-0
:
2014
:
5.1,II
:
34.01 g/mol
:
GHS05
:
GHS07
:
Ambient