

Hydrochloric acid 34 - 37% for trace analysis
KRAF20690.3000
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
:
:110 °C (1013 hPa)
:–30 °C
:1.18 g/cm³ (20 °C)
:8,II
:36.46 g/mol
:231-595-7
:1789
:GHS05
:GHS07
:Ambient
:7647-01-0
:HCl
:MFCD00011324