Order Entry
Germany
ContactUsLinkComponent
Nitric acid 55%
Nitric acid 55%
Catalog # WAKO633-03335
Supplier:  Wako
CAS Number:  
Nitric acid 55%
Catalog # WAKO633-03335
Supplier:  Wako
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349