Order Entry
Germany
ContactUsLinkComponent
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution
Catalog # 662-8355
Supplier:  Mettler - Toledo
CAS Number:  
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution
Catalog # 662-8355
Supplier:  Mettler - Toledo
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

About this item

 :
 :74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl+AgCl
MDL Number:MFCD00011360