Order Entry
Germany
ContactUsLinkComponent
Tween® 80 (Polysorbate) 10% in aqueous solution
Tween® 80 (Polysorbate) 10% in aqueous solution
Catalog # DG013
Supplier:  G-Biosciences
CAS Number:  
Tween® 80 (Polysorbate) 10% in aqueous solution
Catalog # DG013
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    5
  • Seq:
    0005

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:500-019-9
CAS Number:9005-65-6
MDL Number:MFCD00082107