EDTA disodium salt 0.02 mol/l (0.04 N) in aqueous solution for metal titration
Supplier: Bernd Kraft
Certificates
EDTA disodium salt 0.02 mol/l (0.04 N) in aqueous solution for metal titration
Supplier: Bernd Kraft
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals