EDTA disodium salt 0.01783 mol/l (N/28) in aqueous solution for hardness of water determinations
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
Frequently Bought Together