Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
EDTA disodium salt
EDTA disodium salt 0.1 mol/l in aqueous solution
EDTA disodium salt 0.1 mol/l in aqueous solution
EDTA disodium salt 0.1 mol/l in aqueous solution
KRAF01081.3000
:
Bernd Kraft
:
EDTA disodium salt 0.1 mol/l in aqueous solution
KRAF01081.3000
:
Bernd Kraft
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
336.21 g/mol
:
Ambient
:
205-358-3
:
139-33-3
:
C₁₀H₁₄N₂Na₂O₈
:
MFCD00070672