Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-chloroisonicotinate
Methyl-2-chloroisonicotinate
Methyl-2-chloroisonicotinate
Methyl-2-chloroisonicotinate
Supplier:
BIONET RESEARCH
Certificates
Expand image
Methyl-2-chloroisonicotinate
Supplier:
BIONET RESEARCH
About this item
CAS Number:
58481-11-1
:
Formula:
C₇H₆ClNO₂
MDL Number:
MFCD01765409
:
171.5810 g/mol