Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-bromo-3-methylbenzoate
Methyl-4-bromo-3-methylbenzoate
Methyl-4-bromo-3-methylbenzoate
Methyl-4-bromo-3-methylbenzoate
Supplier:
COMBI-BLOCKS
Certificates
Expand image
Methyl-4-bromo-3-methylbenzoate
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
148547-19-7
:
Formula:
C₉H₉BrO₂
MDL Number:
MFCD00673014
:
229.07 g/mol