Order Entry
Germany
ContactUsLinkComponent
Phenolphthalein 0.5% in denatured ethanol 90%
Flammable
Phenolphthalein 0.5% in denatured ethanol 90%
Catalog # KRAF03256.4700
Supplier:  Bernd Kraft
CAS Number:  
undefined
Flammable
Phenolphthalein 0.5% in denatured ethanol 90%
Catalog # KRAF03256.4700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
  • Seq:
    2895

Specifications

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,II
 :GHS02
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913