
Ethylene glycol-[D₄] (98% D), Sigma-Aldrich®
Catalog # SIAL347442-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:66.925 g/mol
:M-Clarity™ quality level = MQ200
EINECS:218-730-5
:GHS07
Storage Temperature:Refrigerator
CAS Number:2219-51-4
Formula:C₂H₆O₂
MDL Number:MFCD00144289