Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-(trifluoromethyl)phenol
3-Bromo-5-(trifluoromethyl)phenol
3-Bromo-5-(trifluoromethyl)phenol
3-Bromo-5-(trifluoromethyl)phenol
Catalog # COBBOT-1171-5G
Supplier:
COMBI-BLOCKS
CAS Number:
Expand image
3-Bromo-5-(trifluoromethyl)phenol
Catalog # COBBOT-1171-5G
Supplier:
COMBI-BLOCKS
CAS Number:
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
:
241.0100 g/mol
CAS Number:
1025718-84-6
Formula:
C₇H₄BrF₃O
MDL Number:
MFCD11041083
Product Details