Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxyisonicotinic acid

2-Hydroxyisonicotinic acid

Marketsource
Supplier:   COMBI-BLOCKS
2-Hydroxyisonicotinic acid
Supplier:  COMBI-BLOCKS
Marketsource
Certificates
2-Hydroxyisonicotinic acid

3 Options Available Below

Product Details & Documents
Synonyms:  2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
  • CAS Number:22282-72-0
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD01318566
  • Molecular Weight:139.11 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource