ContactUsLinkComponent
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Corrosive
Flammable
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
New Product
Certificates
undefined
Corrosive
Flammable
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
New Product
Certificates
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • Water:
    ≤0.5 %
  • Strength:
    1.10 to 1.40 M
  • NMR:
    ≥96.0 %