Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride 5->6 N in 2-propanol, AcroSeal®
Irritant
Corrosive
Flammable
Hydrogen chloride 5->6 N in 2-propanol, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
Hydrogen chloride 5->6 N in 2-propanol, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Water:
    ≤1.0 %
  • Titration with NaOH:
    5 to 6 N

Specification Test Results

Frequently Bought Together