Hydrogen chloride 3 mol/l in 1-butanol, Supelco®
: Merck
Frequently Bought Together
2-Nitrophenylhydrazine, moistened 30% with water 97%
: Thermo Fisher Scientific
Isobutyronitrile 99%
: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, extra pure
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
: PanReac AppliChem
Acetonitrile, anhydrous ≥99.8%
: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Diethyl ether ≥99% stabilised for HPLC
: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals