Order Entry
Germany
Orders LinkContactUsLinkComponent
3,4-Difluoro-2-ethoxybenzonitrile

3,4-Difluoro-2-ethoxybenzonitrile

1
Catalog # CHMPFL13815.POR
Supplier:   CHEMPUR
3,4-Difluoro-2-ethoxybenzonitrile
Catalog # CHMPFL13815.POR
Supplier:  CHEMPUR
CAS Number:  
3,4-Difluoro-2-ethoxybenzonitrile

1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
  • Formula:C₉H₇F₂NO
  • MDL Number:MFCD04973771
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.