Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium chloride
Calcium chloride 30.7 - 31.3% in aqueous solution, extra pure
Calcium chloride 30.7 - 31.3% in aqueous solution, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Calcium chloride 30.7 - 31.3% in aqueous solution, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
10043-52-4
:
Formula:
CaCl₂
MDL Number:
MFCD00010903
:
110.98 g/mol
Storage Temperature:
Ambient
EINECS:
233-140-8