Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Chloromethyl)-3-phenylisoxazole
5-(Chloromethyl)-3-phenylisoxazole
5-(Chloromethyl)-3-phenylisoxazole
5-(Chloromethyl)-3-phenylisoxazole
Supplier:
BIONET RESEARCH
Certificates
Expand image
5-(Chloromethyl)-3-phenylisoxazole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
1011-37-6
:
Formula:
C₁₀H₈ClNO
MDL Number:
MFCD01935949