Order Entry
Germany
ContactUsLinkComponent
Potassium nitrate-[¹⁵N] (60% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL490547-10G
Supplier:  Merck
CAS Number:  
Potassium nitrate-[¹⁵N] (60% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL490547-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:57654-83-8
Formula:KNO₃
MDL Number:MFCD00075049