
2-Iodosobenzoesäure ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:304-91-6
:
Formula:C₇H₅IO₃
MDL Number:MFCD00002401
:264.02 g/mol
EINECS:206-159-4
:GHS07
Storage Temperature:Raumtemperatur
Related Information
Frequently Bought Together