Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-N,N-diisopropyl-3-methylbenzamide
4-Bromo-N,N-diisopropyl-3-methylbenzamide
4-Bromo-N,N-diisopropyl-3-methylbenzamide
4-Bromo-N,N-diisopropyl-3-methylbenzamide
Supplier:
COMBI-BLOCKS
Certificates
Expand image
4-Bromo-N,N-diisopropyl-3-methylbenzamide
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
52010-31-8
:
Formula:
C₁₄H₂₀BrNO
MDL Number:
MFCD09972126