Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-N-cyclohexylbenzenesulphonamide
3-Amino-N-cyclohexylbenzenesulphonamide
3-Amino-N-cyclohexylbenzenesulphonamide
3-Amino-N-cyclohexylbenzenesulphonamide
Supplier:
COMBI-BLOCKS
Certificates
Expand image
3-Amino-N-cyclohexylbenzenesulphonamide
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
61886-26-8
:
Formula:
C₁₂H₁₈N₂O₂S
MDL Number:
MFCD00089429